missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyethyl Salicylate 98.0+%, TCI America™
$32.72 - $163.64
Chemical Identifiers
| CAS | 87-28-5 |
|---|---|
| Molecular Formula | C9H10O4 |
| Molecular Weight (g/mol) | 182.18 |
| MDL Number | MFCD00002862 |
| InChI Key | LVYLCBNXHHHPSB-UHFFFAOYSA-N |
| Synonym | 2-hydroxyethyl salicylate, glycol salicylate, ethylene glycol monosalicylate, espirosal, glycol monosalicylate, rheumacyl, sarocol, spirosal, glysal, phlogont |
| PubChem CID | 6880 |
| ChEBI | CHEBI:86541 |
| IUPAC Name | 2-hydroxyethyl 2-hydroxybenzoate |
| SMILES | OCCOC(=O)C1=CC=CC=C1O |
Chemical Identifiers
| 87-28-5 | |
| 182.18 | |
| LVYLCBNXHHHPSB-UHFFFAOYSA-N | |
| 6880 | |
| 2-hydroxyethyl 2-hydroxybenzoate |
| C9H10O4 | |
| MFCD00002862 | |
| 2-hydroxyethyl salicylate, glycol salicylate, ethylene glycol monosalicylate, espirosal, glycol monosalicylate, rheumacyl, sarocol, spirosal, glysal, phlogont | |
| CHEBI:86541 | |
| OCCOC(=O)C1=CC=CC=C1O |
Specifications
| 87-28-5 | |
| C9H10O4 | |
| 25 g | |
| LVYLCBNXHHHPSB-UHFFFAOYSA-N | |
| 2-hydroxyethyl 2-hydroxybenzoate | |
| 6880 | |
| 182.18 | |
| Crystalline Lumps |
| White | |
| MFCD00002862 | |
| 2-hydroxyethyl salicylate, glycol salicylate, ethylene glycol monosalicylate, espirosal, glycol monosalicylate, rheumacyl, sarocol, spirosal, glysal, phlogont | |
| OCCOC(=O)C1=CC=CC=C1O | |
| 182.18 | |
| CHEBI:86541 | |
| ≥98.0% (GC) | |
| 2-Hydroxyethyl Salicylate |
Safety and Handling
EINECSNumber : (3)-1623&(9)-1496
TSCA : Yes