missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyethyl Salicylate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Boiling Point | 172°C |
| Quantity | 500 g |
| Formula Weight | 182.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 2-Hydroxyethyl Salicylate |
Safety and Handling
| EINECSNumber | (3)-1623&(9)-1496 |
| TSCA | Yes |