missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl Methyl Terephthalate 97.0+%, TCI America™

Catalog No. T16795G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16795G Supplier TCI America Supplier No. T16795G
Only null left

Chemical Identifiers

CAS 3645-00-9
Molecular Formula C11H12O5
Molecular Weight (g/mol) 224.212
MDL Number MFCD01321191
InChI Key DJQMYWWZWUOCBQ-UHFFFAOYSA-N
Synonym Terephthalic Acid 2-Hydroxyethyl Methyl Ester
PubChem CID 77207
IUPAC Name 4-O-(2-hydroxyethyl) 1-O-methyl benzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC=C(C=C1)C(=O)OCCO

Specifications

Melting Point 82°C
Color White
Quantity 5 g
Formula Weight 224.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyethyl Methyl Terephthalate