missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl Methyl Sulfone 98.0+%, TCI America™

Catalog No. H065025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H065025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H065025G Supplier TCI America Supplier No. H065025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15205-66-0
Molecular Formula C3H8O3S
Molecular Weight (g/mol) 124.154
MDL Number MFCD00014747
InChI Key KFTYFTKODBWKOU-UHFFFAOYSA-N
Synonym 2-methylsulfonyl ethanol, 2-hydroxyethyl methyl sulfone, 2-methanesulfonylethanol, 2-methanesulfonylethan-1-ol, 2-methanesulfonyl-ethanol, 2-methanesulfonyl ethanol, ethanol, 2-methylsulfonyl, unii-5fi8eb2k0l, 2-methylsulphonyl ethanol, 2-methylsulfonyl ethyl alcohol
PubChem CID 84834
IUPAC Name 2-methylsulfonylethanol
SMILES CS(=O)(=O)CCO

Specifications

Color White
Boiling Point 149°C
Quantity 25 g
Formula Weight 124.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Hydroxyethyl Methyl Sulfone
EINECSNumber (9)-1419
TSCA No