accessibility menu, dialog, popup

UserName

CAS RN 15205-66-0

IUPAC Name: 2-methanesulfonylethan-1-ol
Synonyms: 2-methanesulfonylethanol
Molecular Weight (g/mol): 124.15
Molecular Formula: C3H8O3S
InChi Key: KFTYFTKODBWKOU-UHFFFAOYSA-N
SMILES: CS(=O)(=O)CCO
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)