missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyethyl Methacrylate (stabilized with MEHQ) 95.0+%, TCI America™
$39.14 - $61.51
Chemical Identifiers
| CAS | 868-77-9 |
|---|---|
| Molecular Formula | C6H10O3 |
| Molecular Weight (g/mol) | 130.143 |
| MDL Number | MFCD00002863 |
| InChI Key | WOBHKFSMXKNTIM-UHFFFAOYSA-N |
| Synonym | 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1 |
| PubChem CID | 13360 |
| ChEBI | CHEBI:34288 |
| IUPAC Name | 2-hydroxyethyl 2-methylprop-2-enoate |
| SMILES | CC(=C)C(=O)OCCO |
Chemical Identifiers
| 868-77-9 | |
| 130.143 | |
| WOBHKFSMXKNTIM-UHFFFAOYSA-N | |
| 13360 | |
| 2-hydroxyethyl 2-methylprop-2-enoate |
| C6H10O3 | |
| MFCD00002863 | |
| 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1 | |
| CHEBI:34288 | |
| CC(=C)C(=O)OCCO |
Specifications
| 868-77-9 | |
| 85°C | |
| MFCD00002863 | |
| 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1 | |
| CC(=C)C(=O)OCCO | |
| 130.143 | |
| CHEBI:34288 | |
| ≥95.0% (GC) | |
| 2-Hydroxyethyl Methacrylate (stabilized with MEHQ) |
| Yellow | |
| C6H10O3 | |
| 25 g | |
| WOBHKFSMXKNTIM-UHFFFAOYSA-N | |
| 2-hydroxyethyl 2-methylprop-2-enoate | |
| 13360 | |
| 130.14 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0958&(2)-1044
RTECSNumber : OZ4725000
TSCA : Yes