missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyethyl Methacrylate (stabilized with MEHQ) 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 85°C |
| Quantity | 25 g |
| Formula Weight | 130.14 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Hydroxyethyl Methacrylate (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (2)-0958&(2)-1044 |
| RTECSNumber | OZ4725000 |
| TSCA | Yes |