Learn More
2-Hydroxyethyl methacrylate, 97%, stab. with 4-methoxyphenol
CAS: 868-77-9 | C6H10O3 | 130.143 g/mol
$49.42 - $127.99
Chemical Identifiers
| CAS | 868-77-9 |
|---|---|
| Molecular Formula | C6H10O3 |
| Molecular Weight (g/mol) | 130.143 |
| MDL Number | MFCD00002863 |
| InChI Key | WOBHKFSMXKNTIM-UHFFFAOYSA-N |
| Synonym | 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1 |
| PubChem CID | 13360 |
| ChEBI | CHEBI:34288 |
| IUPAC Name | 2-hydroxyethyl 2-methylprop-2-enoate |
| SMILES | CC(=C)C(=O)OCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2426022
|
Thermo Scientific Chemicals
B2426022 |
100 g |
Each for $49.42
|
|
|||||
|
AAB2426036
|
Thermo Scientific Chemicals
B2426036 |
500 g |
Each for $127.99
|
|
|||||
Description
2-Hydroxyethyl methacrylate is used in UV-curable inks and coatings. It is also used in adhesives, artificial nails, dental materials and lacquers. In dentistry, it is one of the main volatile acrylates along with methyl methacrylate. Further, it is used as a monomer in the synthesis of polymers for dental prosthetics and for geotechnical grouting in construction work.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxyethyl methacrylate is used in UV-curable inks and coatings. It is also used in adhesives, artificial nails, dental materials and lacquers. In dentistry, it is one of the main volatile acrylates along with methyl methacrylate. Further, it is used as a monomer in the synthesis of polymers for dental prosthetics and for geotechnical grouting in construction work.
Solubility
Slightly miscible with hot water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 868-77-9 | |
| 130.143 | |
| WOBHKFSMXKNTIM-UHFFFAOYSA-N | |
| 13360 | |
| 2-hydroxyethyl 2-methylprop-2-enoate |
| C6H10O3 | |
| MFCD00002863 | |
| 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1 | |
| CHEBI:34288 | |
| CC(=C)C(=O)OCCO |
Specifications
| 868-77-9 | |
| 1.071 | |
| 101°C (213°F) | |
| C6H10O3 | |
| MFCD00002863 | |
| 1071583 | |
| 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1 | |
| WOBHKFSMXKNTIM-UHFFFAOYSA-N | |
| 2-hydroxyethyl 2-methylprop-2-enoate | |
| 13360 | |
| 130.14 | |
| 2-Hydroxyethyl methacrylate, stabilized with ≈500ppm 4-methoxyphenol |
| -12°C | |
| 250°C | |
| Ester-like | |
| 1.453 | |
| 100 g | |
| Light sensitive | |
| Slightly miscible with hot water. | |
| CC(=C)C(=O)OCCO | |
| 130.143 | |
| CHEBI:34288 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
H315-H317-H319
EINECSNumber : 212-782-2
RTECSNumber : OZ4725000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only