missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl methacrylate, 97%, stab. with 4-methoxyphenol

Catalog No. AAB2426022
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2426022 100 g
AAB2426036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2426022 Supplier Thermo Scientific Chemicals Supplier No. B2426022

CAS: 868-77-9 | C6H10O3 | 130.143 g/mol

2-Hydroxyethyl methacrylate is used in UV-curable inks and coatings. It is also used in adhesives, artificial nails, dental materials and lacquers. In dentistry, it is one of the main volatile acrylates along with methyl methacrylate. Further, it is used as a monomer in the synthesis of polymers for dental prosthetics and for geotechnical grouting in construction work.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxyethyl methacrylate is used in UV-curable inks and coatings. It is also used in adhesives, artificial nails, dental materials and lacquers. In dentistry, it is one of the main volatile acrylates along with methyl methacrylate. Further, it is used as a monomer in the synthesis of polymers for dental prosthetics and for geotechnical grouting in construction work.

Solubility
Slightly miscible with hot water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 868-77-9
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00002863
InChI Key WOBHKFSMXKNTIM-UHFFFAOYSA-N
Synonym 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1
PubChem CID 13360
ChEBI CHEBI:34288
IUPAC Name 2-hydroxyethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCO

Specifications

Melting Point -12°C
Density 1.071
Boiling Point 250°C
Flash Point 101°C (213°F)
Odor Ester-like
Refractive Index 1.453
Quantity 100 g
Beilstein 1071583
Sensitivity Light sensitive
Solubility Information Slightly miscible with hot water.
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material 2-Hydroxyethyl methacrylate, stabilized with ≈500ppm 4-methoxyphenol
Show More Show Less
Hazard Category H315-H317-H319
Hazard Statement GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
EINECSNumber 212-782-2
RTECSNumber OZ4725000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only