missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyethyl Acetate 60.0+%, TCI America™
$158.44 - $704.62
Chemical Identifiers
| CAS | 542-59-6 |
|---|---|
| Molecular Formula | C4H8O3 |
| Molecular Weight (g/mol) | 104.105 |
| MDL Number | MFCD00002864 |
| InChI Key | HXDLWJWIAHWIKI-UHFFFAOYSA-N |
| Synonym | Ethylene Glycol Monoacetate, Acetic Acid 2-Hydroxyethyl Ester |
| PubChem CID | 10960 |
| IUPAC Name | 2-hydroxyethyl acetate |
| SMILES | CC(=O)OCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A105925ML
|
TCI America
A105925ML |
25 mL |
Each for $158.44
|
|
|||||
|
A1059500ML
|
TCI America
A1059500ML |
500 mL |
Each for $704.62
|
|
|||||
Chemical Identifiers
| 542-59-6 | |
| 104.105 | |
| HXDLWJWIAHWIKI-UHFFFAOYSA-N | |
| 10960 | |
| CC(=O)OCCO |
| C4H8O3 | |
| MFCD00002864 | |
| Ethylene Glycol Monoacetate, Acetic Acid 2-Hydroxyethyl Ester | |
| 2-hydroxyethyl acetate |
Specifications
| 542-59-6 | |
| 182°C | |
| MFCD00002864 | |
| Ethylene Glycol Monoacetate, Acetic Acid 2-Hydroxyethyl Ester | |
| CC(=O)OCCO | |
| 104.105 | |
| 104.11 | |
| Liquid |
| Colorless | |
| C4H8O3 | |
| 25 mL | |
| HXDLWJWIAHWIKI-UHFFFAOYSA-N | |
| 2-hydroxyethyl acetate | |
| 10960 | |
| ≥60.0% (GC) | |
| 2-Hydroxyethyl Acetate |
Safety and Handling
EINECSNumber : (2)-0738&(2)-0665
RTECSNumber : KW7175000
TSCA : Yes