CAS RN 542-59-6
CAS RN 542-59-6
IUPAC Name:
2-hydroxyethyl acetate
Synonyms:
laneth-9 acetate laneth-10 acetate
Molecular Weight (g/mol):
104.11
Molecular Formula:
C4H8O3
InChi Key:
HXDLWJWIAHWIKI-UHFFFAOYSA-N
SMILES:
CC(=O)OCCO
1
–
1
of
1
results
1
2-Hydroxyethyl Acetate 60.0+%, TCI America™
CAS: 542-59-6 Molecular Formula: C4H8O3 Molecular Weight (g/mol): 104.105 MDL Number: MFCD00002864 InChI Key: HXDLWJWIAHWIKI-UHFFFAOYSA-N Synonym: Ethylene Glycol Monoacetate, Acetic Acid 2-Hydroxyethyl Ester PubChem CID: 10960 IUPAC Name: 2-hydroxyethyl acetate SMILES: CC(=O)OCCO