missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxycyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7131028
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14091G 1 g
H14095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14091G Supplier TCI America Supplier No. H14091G
Only null left

Chemical Identifiers

CAS 609-69-8
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD02684437
InChI Key SNKAANHOVFZAMR-UHFFFAOYNA-N
PubChem CID 271816
ChEBI CHEBI:37377
IUPAC Name 2-hydroxycyclohexane-1-carboxylic acid
SMILES OC1CCCCC1C(O)=O

Specifications

Melting Point 69°C
Color White
Quantity 1 g
Formula Weight 144.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxycyclohexanecarboxylic Acid (cis- and trans- mixture)