accessibility menu, dialog, popup

UserName

OOHHO

CAS RN 609-69-8

IUPAC Name: 2-hydroxycyclohexane-1-carboxylic acid
Synonyms: 2-hydroxycyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: SNKAANHOVFZAMR-UHFFFAOYNA-N
SMILES: OC1CCCCC1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results