missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxychalcone 98.0+%, TCI America™
Supplier: TCI America H023425G
Specifications
| 2-Hydroxychalcone | |
| C15H12O2 | |
| 25 g | |
| UDOOPSJCRMKSGL-ZHACJKMWSA-N | |
| (E)-3-(2-hydroxyphenyl)-1-phenylprop-2-en-1-one | |
| 5367146 | |
| ≥98.0% (GC,T) |
| 644-78-0 | |
| MFCD00016449 | |
| 2-hydroxychalcone, 2e-3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 2-propen-1-one, 3-2-hydroxyphenyl-1-phenyl, 2-2-hydroxybenzal acetophenone, e-3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 3-2-hydroxyphenyl-1-phenyl-2-propen-1-one, e-3-2-hydroxyphenyl-1-phenyl-prop-2-en-1-one, 2-hydroxybenzylidene acetophenone, chalcone, 1 | |
| C1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2O | |
| 224.259 | |
| 224.26 | |
| Crystalline Powder |
Chemical Identifiers
| 644-78-0 | |
| 224.259 | |
| UDOOPSJCRMKSGL-ZHACJKMWSA-N | |
| 5367146 | |
| C1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2O |
| C15H12O2 | |
| MFCD00016449 | |
| 2-hydroxychalcone, 2e-3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 2-propen-1-one, 3-2-hydroxyphenyl-1-phenyl, 2-2-hydroxybenzal acetophenone, e-3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 3-2-hydroxyphenyl-1-phenyl-2-propen-1-one, e-3-2-hydroxyphenyl-1-phenyl-prop-2-en-1-one, 2-hydroxybenzylidene acetophenone, chalcone, 1 | |
| (E)-3-(2-hydroxyphenyl)-1-phenylprop-2-en-1-one |
Safety and Handling
TSCA : No