accessibility menu, dialog, popup

UserName

OHO(E)

CAS RN 644-78-0

IUPAC Name: (2E)-3-(2-hydroxyphenyl)-1-phenylprop-2-en-1-one
Synonyms: (2E)-3-(2-hydroxyphenyl)-1-phenylprop-2-en-1-one
Molecular Weight (g/mol): 224.26
Molecular Formula: C15H12O2
InChi Key: UDOOPSJCRMKSGL-ZHACJKMWSA-N
SMILES: OC1=CC=CC=C1\C=C\C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)