missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxychalcone 98.0+%, TCI America™

Catalog No. H02345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02345G 5 g
H023425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H02345G Supplier TCI America Supplier No. H02345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 644-78-0
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00016449
InChI Key UDOOPSJCRMKSGL-ZHACJKMWSA-N
Synonym 2-hydroxychalcone, 2e-3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 2-propen-1-one, 3-2-hydroxyphenyl-1-phenyl, 2-2-hydroxybenzal acetophenone, e-3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 3-2-hydroxyphenyl-1-phenyl-2-propen-1-one, e-3-2-hydroxyphenyl-1-phenyl-prop-2-en-1-one, 2-hydroxybenzylidene acetophenone, chalcone, 1
PubChem CID 5367146
IUPAC Name (E)-3-(2-hydroxyphenyl)-1-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2O

Specifications

Quantity 5 g
Formula Weight 224.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxychalcone