missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybicyclo[3.2.1]octane-6-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTBG00035DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. BTBG00035DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

257932-29-9
170.21
FSQSHXLJRFYYMV-UHFFFAOYNA-N
2802922
OC1CCC2CC1CC2C(O)=O
C9H14O3
MFCD00180256
2-hydroxybicyclo 3.2.1 octane-6-carboxylic acid, maybridge1_002398, 4-hydroxybicyclo 3.2.1 octane-7-carboxylic acid, bicyclo 3.2.1 octane-6-carboxylicacid, 2-hydroxy
4-hydroxybicyclo[3.2.1]octane-7-carboxylic acid

Specifications

2-Hydroxybicyclo[3.2.1]octane-6-carboxylic acid
96.5
97%
C9H14O3
MFCD00180256
FSQSHXLJRFYYMV-UHFFFAOYNA-N
4-hydroxybicyclo[3.2.1]octane-7-carboxylic acid
2802922
97%
257932-29-9
100.0
Amber Glass Bottle
1g
2-hydroxybicyclo 3.2.1 octane-6-carboxylic acid, maybridge1_002398, 4-hydroxybicyclo 3.2.1 octane-7-carboxylic acid, bicyclo 3.2.1 octane-6-carboxylicacid, 2-hydroxy
OC1CCC2CC1CC2C(O)=O
170.21
170.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions