missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybicyclo[3.2.1]octane-6-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-3110273
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTBG00035CB 250mg Amber Glass Bottle
BTBG00035DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTBG00035CB Supplier Thermo Scientific Chemicals Supplier No. BTBG00035CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 257932-29-9
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00180256
InChI Key FSQSHXLJRFYYMV-UHFFFAOYNA-N
Synonym 2-hydroxybicyclo 3.2.1 octane-6-carboxylic acid, maybridge1_002398, 4-hydroxybicyclo 3.2.1 octane-7-carboxylic acid, bicyclo 3.2.1 octane-6-carboxylicacid, 2-hydroxy
PubChem CID 2802922
IUPAC Name 4-hydroxybicyclo[3.2.1]octane-7-carboxylic acid
SMILES OC1CCC2CC1CC2C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 170.21
Percent Purity 97%
Chemical Name or Material 2-Hydroxybicyclo[3.2.1]octane-6-carboxylic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.