Learn More
2-Hydroxybenzylamine, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 932-30-9 |
|---|---|
| Molecular Formula | C7H9NO |
| Molecular Weight (g/mol) | 123.15 |
| MDL Number | MFCD00870498 |
| InChI Key | KPRZOPQOBJRYSW-UHFFFAOYSA-N |
| Synonym | 2-aminomethyl phenol, 2-hydroxybenzylamine, phenol, 2-aminomethyl, o-hydroxybenzylamine, 2-hydroxyphenyl methylamine, unii-696r5n4nrm, salicylamine, aminomethyl phenol, o-aminomethylphenol, o-hydroxy-benzylamine |
| PubChem CID | 70267 |
| IUPAC Name | 2-(aminomethyl)phenol |
| SMILES | C1=CC=C(C(=C1)CN)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC298890010
|
Thermo Scientific Chemicals
298890010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC298890050
|
Thermo Scientific Chemicals
298890050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 932-30-9 | |
| 123.15 | |
| KPRZOPQOBJRYSW-UHFFFAOYSA-N | |
| 70267 | |
| C1=CC=C(C(=C1)CN)O |
| C7H9NO | |
| MFCD00870498 | |
| 2-aminomethyl phenol, 2-hydroxybenzylamine, phenol, 2-aminomethyl, o-hydroxybenzylamine, 2-hydroxyphenyl methylamine, unii-696r5n4nrm, salicylamine, aminomethyl phenol, o-aminomethylphenol, o-hydroxy-benzylamine | |
| 2-(aminomethyl)phenol |
Specifications
| 932-30-9 | |
| 100.0 | |
| Glass bottle | |
| OHC6H4CH2NH2 | |
| 1g | |
| KPRZOPQOBJRYSW-UHFFFAOYSA-N | |
| 2-(aminomethyl)phenol | |
| 70267 | |
| 98% |
| 97.5 | |
| 97.5% min. (GC) | |
| C7H9NO | |
| MFCD00870498 | |
| 2-aminomethyl phenol, 2-hydroxybenzylamine, phenol, 2-aminomethyl, o-hydroxybenzylamine, 2-hydroxyphenyl methylamine, unii-696r5n4nrm, salicylamine, aminomethyl phenol, o-aminomethylphenol, o-hydroxy-benzylamine | |
| C1=CC=C(C(=C1)CN)O | |
| 123.15 | |
| 123.15 | |
| 2-Hydroxybenzylamine, 98% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 213-249-7
RUO â Research Use Only