accessibility menu, dialog, popup

UserName

H2NHO

CAS RN 932-30-9

IUPAC Name: 2-(aminomethyl)phenol
Synonyms: 2-(aminomethyl)phenol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: KPRZOPQOBJRYSW-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results