missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxybenzyl Alcohol 98.0+%, TCI America™
$173.37 - $1288.06
Chemical Identifiers
| CAS | 90-01-7 |
|---|---|
| Molecular Formula | C7H8O2 |
| Molecular Weight (g/mol) | 124.14 |
| MDL Number | MFCD00004617 |
| InChI Key | CQRYARSYNCAZFO-UHFFFAOYSA-N |
| Synonym | salicyl alcohol, 2-hydroxybenzyl alcohol, saligenin, 2-hydroxymethyl phenol, 2-methylolphenol, o-hydroxybenzyl alcohol, saligenol, benzenemethanol, 2-hydroxy, diathesin, o-methylolphenol |
| PubChem CID | 5146 |
| ChEBI | CHEBI:16464 |
| IUPAC Name | 2-(hydroxymethyl)phenol |
| SMILES | OCC1=CC=CC=C1O |
Chemical Identifiers
| 90-01-7 | |
| 124.14 | |
| CQRYARSYNCAZFO-UHFFFAOYSA-N | |
| 5146 | |
| 2-(hydroxymethyl)phenol |
| C7H8O2 | |
| MFCD00004617 | |
| salicyl alcohol, 2-hydroxybenzyl alcohol, saligenin, 2-hydroxymethyl phenol, 2-methylolphenol, o-hydroxybenzyl alcohol, saligenol, benzenemethanol, 2-hydroxy, diathesin, o-methylolphenol | |
| CHEBI:16464 | |
| OCC1=CC=CC=C1O |
Specifications
| 90-01-7 | |
| C7H8O2 | |
| 25 g | |
| CQRYARSYNCAZFO-UHFFFAOYSA-N | |
| 2-(hydroxymethyl)phenol | |
| 5146 | |
| 124.14 | |
| Crystalline Powder |
| 86°C | |
| MFCD00004617 | |
| salicyl alcohol, 2-hydroxybenzyl alcohol, saligenin, 2-hydroxymethyl phenol, 2-methylolphenol, o-hydroxybenzyl alcohol, saligenol, benzenemethanol, 2-hydroxy, diathesin, o-methylolphenol | |
| OCC1=CC=CC=C1O | |
| 124.14 | |
| CHEBI:16464 | |
| ≥98.0% (GC) | |
| 2-Hydroxybenzyl Alcohol |
Safety and Handling
EINECSNumber : (3)-1052
RTECSNumber : DO6430000
TSCA : Yes