missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzyl Alcohol 98.0+%, TCI America™

Catalog No. H022325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H022325G 25 g
H0223250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H022325G Supplier TCI America Supplier No. H022325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90-01-7
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00004617
InChI Key CQRYARSYNCAZFO-UHFFFAOYSA-N
Synonym salicyl alcohol, 2-hydroxybenzyl alcohol, saligenin, 2-hydroxymethyl phenol, 2-methylolphenol, o-hydroxybenzyl alcohol, saligenol, benzenemethanol, 2-hydroxy, diathesin, o-methylolphenol
PubChem CID 5146
ChEBI CHEBI:16464
IUPAC Name 2-(hydroxymethyl)phenol
SMILES OCC1=CC=CC=C1O

Specifications

Melting Point 86°C
Quantity 25 g
Formula Weight 124.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxybenzyl Alcohol
EINECSNumber (3)-1052
RTECSNumber DO6430000
TSCA Yes