Learn More
2-Hydroxybenzyl alcohol, 99%
CAS: 90-01-7 | C7H8O2 | 124.14 g/mol
$111.38 - $1551.70
Chemical Identifiers
| CAS | 90-01-7 |
|---|---|
| Molecular Formula | C7H8O2 |
| Molecular Weight (g/mol) | 124.14 |
| MDL Number | MFCD00004617 |
| InChI Key | CQRYARSYNCAZFO-UHFFFAOYSA-N |
| Synonym | salicyl alcohol, 2-hydroxybenzyl alcohol, saligenin, 2-hydroxymethyl phenol, 2-methylolphenol, o-hydroxybenzyl alcohol, saligenol, benzenemethanol, 2-hydroxy, diathesin, o-methylolphenol |
| PubChem CID | 5146 |
| ChEBI | CHEBI:16464 |
| IUPAC Name | 2-(hydroxymethyl)phenol |
| SMILES | OCC1=CC=CC=C1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1441018
|
Thermo Scientific Chemicals
A1441018 |
50 g |
Each for $111.38
|
|
|||||
|
AAA1441030
|
Thermo Scientific Chemicals
A1441030 |
250 g |
Each for $427.21
|
|
|||||
|
AAA144100B
|
Thermo Scientific Chemicals
A144100B |
1000 g |
Each for $1,551.70
|
|
|||||
Description
2-Hydroxybenzyl alcohol acts as a local anesthetic. It is used in gastrodin production via biotransformation by cultured cells of Aspergillus foetidus and Penicillium cyclopium and participates in the selective ring opening reaction of 4H-1,3,2-benzodioxasilines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxybenzyl alcohol acts as a local anesthetic. It is used in gastrodin production via biotransformation by cultured cells of Aspergillus foetidus and Penicillium cyclopium and participates in the selective ring opening reaction of 4H-1,3,2-benzodioxasilines.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides and strong oxidizing agents.
Chemical Identifiers
| 90-01-7 | |
| 124.14 | |
| CQRYARSYNCAZFO-UHFFFAOYSA-N | |
| 5146 | |
| 2-(hydroxymethyl)phenol |
| C7H8O2 | |
| MFCD00004617 | |
| salicyl alcohol, 2-hydroxybenzyl alcohol, saligenin, 2-hydroxymethyl phenol, 2-methylolphenol, o-hydroxybenzyl alcohol, saligenol, benzenemethanol, 2-hydroxy, diathesin, o-methylolphenol | |
| CHEBI:16464 | |
| OCC1=CC=CC=C1O |
Specifications
| 90-01-7 | |
| C7H8O2 | |
| 50 g | |
| 14,8325 | |
| Soluble in water. | |
| OCC1=CC=CC=C1O | |
| 124.14 | |
| CHEBI:16464 | |
| 99% |
| 83°C to 87°C | |
| MFCD00004617 | |
| 1907195 | |
| salicyl alcohol, 2-hydroxybenzyl alcohol, saligenin, 2-hydroxymethyl phenol, 2-methylolphenol, o-hydroxybenzyl alcohol, saligenol, benzenemethanol, 2-hydroxy, diathesin, o-methylolphenol | |
| CQRYARSYNCAZFO-UHFFFAOYSA-N | |
| 2-(hydroxymethyl)phenol | |
| 5146 | |
| 124.14 | |
| 2-Hydroxybenzyl alcohol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 201-960-5
RTECSNumber : DO6430000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only