missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzotrifluoride 98.0+%, TCI America™

Catalog No. H064710G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H064710G 10 g
H064725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H064710G Supplier TCI America Supplier No. H064710G

Chemical Identifiers

CAS 444-30-4
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.111
MDL Number MFCD00002222
InChI Key ZOQOPXVJANRGJZ-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenol, 2-hydroxybenzotrifluoride, o-trifluoromethylphenol, o-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-o-cresol, phenol, 2-trifluoromethyl, o-trifluoromethoxy phenol, a,a,a-trifluoro-o-cresol, 2-hydroxy-alpha,alpha,alpha-trifluorotoluene
PubChem CID 67958
IUPAC Name 2-(trifluoromethyl)phenol
SMILES C1=CC=C(C(=C1)C(F)(F)F)O

Specifications

Melting Point 45°C
Color White
Boiling Point 149°C
Quantity 10 g
Formula Weight 162.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxybenzotrifluoride