missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzonitrile, 98%

Catalog No. p-7021322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1360606 5 g
AAA1360614 25 g
AAA1360622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1360606 Supplier Thermo Scientific Chemicals Supplier No. A1360606
Only null left

CAS: 611-20-1 | C7H5NO | 119.123 g/mol

2-Hydroxybenzonitrile is used as starting reagent during the synthesis of mono-alkoxyphenyloxazoline, furanoside.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzonitrile is used as starting reagent during the synthesis of mono-alkoxyphenyloxazoline, furanoside.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 611-20-1
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.123
MDL Number MFCD00002145
InChI Key CHZCERSEMVWNHL-UHFFFAOYSA-N
Synonym 2-cyanophenol, o-cyanophenol, salicylonitrile, o-hydroxybenzonitrile, benzonitrile, 2-hydroxy, salicylnitrile, benzonitrile, hydroxy, benzonitrile, o-hydroxy, o-hydoxybenzonitrile, 2-hydroxy-benzonitrile
PubChem CID 11907
IUPAC Name 2-hydroxybenzonitrile
SMILES C1=CC=C(C(=C1)C#N)O

Specifications

Melting Point 92°C to 95°C
Boiling Point 149°C (14 mmHg)
Quantity 5 g
Beilstein 1210029
Solubility Information Soluble in water.
Formula Weight 119.12
Percent Purity 98%
Chemical Name or Material 2-Hydroxybenzonitrile
Hazard Category H302-H317-H318
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c
EINECSNumber 210-259-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only