missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzhydrazide, 98+%

Catalog No. AAA1421814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1421814 25 g
AAA1421822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1421814 Supplier Thermo Scientific Chemicals Supplier No. A1421814
Only null left

CAS: 936-02-7 | C7H8N2O2 | 152.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 936-02-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007599
InChI Key XSXYESVZDBAKKT-UHFFFAOYSA-N
Synonym salicylhydrazide, 2-hydroxybenzhydrazide, salicyl hydrazide, salicyloyl hydrazide, salicylic acid hydrazide, salicylic hydrazide, salicyloylhydrazine, salicylohydrazide, o-hydroxybenzhydrazide, o-hydroxybenzoylhydrazide
PubChem CID 13637
IUPAC Name 2-hydroxybenzohydrazide
SMILES C1=CC=C(C(=C1)C(=O)NN)O

Specifications

Melting Point 146°C to 149°C
Quantity 25 g
Beilstein 472355
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxybenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-311-3
RTECSNumber VO3700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only