missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzeneboronic acid, 97%

Catalog No. p-7058507
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1940003 1 g
AAL1940006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1940003 Supplier Thermo Scientific Chemicals Supplier No. L1940003
Only null left

CAS: 89466-08-0 | C6H7BO3 | 137.93 g/mol

2-Hydroxybenzeneboronic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzeneboronic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 89466-08-0
Molecular Formula C6H7BO3
Molecular Weight (g/mol) 137.93
MDL Number MFCD01074581
InChI Key YDMRDHQUQIVWBE-UHFFFAOYSA-N
Synonym 2-hydroxyphenyl boronic acid, 2-hydroxybenzeneboronic acid, o-hydroxyphenylboronic acid, 2-boronophenol, 2-hydroxyphenyl boranediol, 2-hydroxyphenylboronicacid, boronic acid, 2-hydroxyphenyl, pubchem9522, 2-hydroxyphenylboronic, acmc-209r1d
PubChem CID 2773454
IUPAC Name (2-hydroxyphenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1O

Specifications

Melting Point ∼185°C
Quantity 1 g
Beilstein 2962938
Solubility Information Insoluble in water.
Formula Weight 137.93
Percent Purity 97%
Chemical Name or Material 2-Hydroxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only