missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxyacetanilide 99.0+%, TCI America™

Catalog No. H018925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H018925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H018925G Supplier TCI America Supplier No. H018925G

Chemical Identifiers

CAS 614-80-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002181
InChI Key ADVGKWPZRIDURE-UHFFFAOYSA-N
Synonym 2-acetamidophenol, n-2-hydroxyphenyl acetamide, 2'-hydroxyacetanilide, o-hydroxyacetanilide, 2-acetaminophenol, 2-hydroxyacetanilide, o-acetamidophenol, o-acetylamino phenol, 2-acetylamino phenol, o-acetaminophenol
PubChem CID 11972
IUPAC Name N-(2-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1O

Specifications

Melting Point 209°C
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxyacetanilide
EINECSNumber (3)-0678&(3)-2794
RTECSNumber AE4025000
TSCA Yes