Learn More
Thermo Scientific Chemicals 2-Hydroxy-p-naphthoquinone, 99%
CAS: 83-72-7 | C10H6O3 | 174.16 g/mol
$92.18 - $92.18
Chemical Identifiers
| CAS | 83-72-7 |
|---|---|
| Molecular Formula | C10H6O3 |
| Molecular Weight (g/mol) | 174.16 |
| MDL Number | MFCD00001678 |
| InChI Key | WVCHIGAIXREVNS-UHFFFAOYSA-N |
| Synonym | lawsone, 2-hydroxy-1,4-naphthoquinone, 2-hydroxynaphthalene-1,4-dione, 2-hydroxynaphthoquinone, henna, mehendi, hana, lawson, mendi, flower of paradise |
| PubChem CID | 6755 |
| IUPAC Name | 4-hydroxynaphthalene-1,2-dione |
| SMILES | C1=CC=C2C(=C1)C(=CC(=O)C2=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC121630250
|
Thermo Scientific Chemicals
121630250 |
25 g | Glass bottle |
Each for $92.18
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 83-72-7 | |
| 174.16 | |
| WVCHIGAIXREVNS-UHFFFAOYSA-N | |
| 6755 | |
| C1=CC=C2C(=C1)C(=CC(=O)C2=O)O |
| C10H6O3 | |
| MFCD00001678 | |
| lawsone, 2-hydroxy-1,4-naphthoquinone, 2-hydroxynaphthalene-1,4-dione, 2-hydroxynaphthoquinone, henna, mehendi, hana, lawson, mendi, flower of paradise | |
| 4-hydroxynaphthalene-1,2-dione |
Specifications
| 83-72-7 | |
| 100.0 | |
| Yellow | |
| 98.5% min. (HPLC) | |
| C10H6O3 | |
| 25 g | |
| 01,484 | |
| lawsone, 2-hydroxy-1,4-naphthoquinone, 2-hydroxynaphthalene-1,4-dione, 2-hydroxynaphthoquinone, henna, mehendi, hana, lawson, mendi, flower of paradise | |
| WVCHIGAIXREVNS-UHFFFAOYSA-N | |
| 4-hydroxynaphthalene-1,2-dione | |
| 6755 | |
| 99% | |
| Crystalline Powder |
| 98.5 | |
| 192.0°C to 195.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00001678 | |
| 08, 300 | |
| 15, 5448 | |
| Solubility in water: 2g/L water (20°C). Other solubilities: soluble in benzene,carbon tetrachloride,aceton,,petroleum ether and sodium hydroxide | |
| C1=CC=C2C(=C1)C(=CC(=O)C2=O)O | |
| 174.16 | |
| 174.16 | |
| Pure | |
| 2-Hydroxy-p-naphthoquinone, 99% |
Safety and Handling
GHS H Statement
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get
GHS Signal Word: Warning
EINECSNumber : 201-496-3
RTECSNumber : QL8200000
TSCA : TSCA
RUO – Research Use Only