missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-n-octanoic Acid 98.0+%, TCI America™

Catalog No. H05925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05925G 5 g
H059225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05925G Supplier TCI America Supplier No. H05925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 617-73-2
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.213
MDL Number MFCD00014410
InChI Key JKRDADVRIYVCCY-UHFFFAOYSA-N
Synonym 2-hydroxycaprylic acid, hydroxyoctanoate, hydroxycaprylic acid, 2-hydroxy caprylic acid, 2-hydroxy-octanoic acid, octanoic acid, hydroxy, alpha-hydroxycaprylic acid, 2-hydroxy-n-octanoic acid, 2-hydroxyoctanoate, alpha-hydroxyoctanoic acid
PubChem CID 94180
ChEBI CHEBI:86543
IUPAC Name 2-hydroxyoctanoic acid
SMILES CCCCCCC(C(=O)O)O

Specifications

Melting Point 69°C
Color White
Quantity 5 g
Formula Weight 160.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-n-octanoic Acid
EINECSNumber (2)-1340
TSCA No