missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-9-fluorenone 90.0+%, TCI America™

Catalog No. H03291G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H03291G Supplier TCI America Supplier No. H03291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6949-73-1
Molecular Formula C13H8O2
Molecular Weight (g/mol) 196.205
MDL Number MFCD00001155
InChI Key GXUBPHMYNSICJC-UHFFFAOYSA-N
Synonym 2-hydroxy-9-fluorenone, 2-hydroxy-9h-fluoren-9-one, 2-hydroxy-fluoren-9-one, acmc-209o8z, 9h-fluoren-9-one,2-hydroxy, 2-hydroxy-9h-fluoren-9-one #, 9h-fluoren-9-one, 2-hydroxy
PubChem CID 81386
IUPAC Name 2-hydroxyfluoren-9-one
SMILES C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)O

Specifications

Melting Point 205°C
Quantity 1 g
Formula Weight 196.21
Percent Purity ≥90.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-9-fluorenone