missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-6-methylpyridine 98.0+%, TCI America™
Supplier: TCI America H094225G
Specifications
| 2-Hydroxy-6-methylpyridine | |
| 159°C | |
| MFCD00006279 | |
| 2-hydroxy-6-methylpyridine, 6-methylpyridin-2-ol, 6-methylpyridin-2 1h-one, 6-methyl-2-pyridinol, 6-methyl-2-pyridone, 6-methyl-2-pyridinone, 2 1h-pyridinone, 6-methyl, 6-methyl-2 1h-pyridinone, 2-hydroxy-6-picoline, 6-hydroxy-2-picoline | |
| CC1=CC=CC(=O)N1 | |
| 109.13 | |
| 109.13 | |
| Crystalline Powder |
| 3279-76-3 | |
| C6H7NO | |
| 25 g | |
| JEAVIRYCMBDJIU-UHFFFAOYSA-N | |
| 6-methyl-1,2-dihydropyridin-2-one | |
| 76772 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 3279-76-3 | |
| 109.13 | |
| JEAVIRYCMBDJIU-UHFFFAOYSA-N | |
| 76772 | |
| CC1=CC=CC(=O)N1 |
| C6H7NO | |
| MFCD00006279 | |
| 2-hydroxy-6-methylpyridine, 6-methylpyridin-2-ol, 6-methylpyridin-2 1h-one, 6-methyl-2-pyridinol, 6-methyl-2-pyridone, 6-methyl-2-pyridinone, 2 1h-pyridinone, 6-methyl, 6-methyl-2 1h-pyridinone, 2-hydroxy-6-picoline, 6-hydroxy-2-picoline | |
| 6-methyl-1,2-dihydropyridin-2-one |
Safety and Handling
TSCA : No