missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methylpyridine 98.0+%, TCI America™

Catalog No. p-7130818
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09425G 5 g
H094225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09425G Supplier TCI America Supplier No. H09425G
Only null left

Chemical Identifiers

CAS 3279-76-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006279
InChI Key JEAVIRYCMBDJIU-UHFFFAOYSA-N
Synonym 2-hydroxy-6-methylpyridine, 6-methylpyridin-2-ol, 6-methylpyridin-2 1h-one, 6-methyl-2-pyridinol, 6-methyl-2-pyridone, 6-methyl-2-pyridinone, 2 1h-pyridinone, 6-methyl, 6-methyl-2 1h-pyridinone, 2-hydroxy-6-picoline, 6-hydroxy-2-picoline
PubChem CID 76772
IUPAC Name 6-methyl-1,2-dihydropyridin-2-one
SMILES CC1=CC=CC(=O)N1

Specifications

Melting Point 159°C
Quantity 5 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-6-methylpyridine