missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methylnicotinic Acid 98.0+%, TCI America™

Catalog No. p-7130710
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
H07575G 5 g
H075725G 25 g
2 options

Catalog No. H07575G

Supplier: TCI America H07575G

Only null left

Chemical Identifiers

CAS 38116-61-9
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00006272
InChI Key XRIHTJYXIHOBDQ-UHFFFAOYSA-N
Synonym 2-hydroxy-6-methylnicotinic acid, 2-hydroxy-6-methylpyridine-3-carboxylic acid, 6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid, chembl50725, 3-pyridinecarboxylic acid, 1,2-dihydro-6-methyl-2-oxo, 2-hydroxy-6-methyl nicotinic acid, 2-hydroxy-6-methyl-nicotinic acid, 6-methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid, 6-methyl-2-oxohydropyridine-3-carboxylic acid, 2-hydroxy-6-methylnicotinicacid
PubChem CID 94960
IUPAC Name 6-methyl-2-oxo-1H-pyridine-3-carboxylic acid
SMILES CC1=CC=C(C(=O)N1)C(=O)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 153.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-6-methylnicotinic Acid