accessibility menu, dialog, popup

UserName

CAS RN 38116-61-9

CH3OOOHN

CAS RN 38116-61-9

IUPAC Name: 6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate
Synonyms: 6-methyl-2-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 152.13
Molecular Formula: C7H6NO3
InChi Key: XRIHTJYXIHOBDQ-UHFFFAOYSA-M
SMILES: CC1=CC=C(C([O-])=O)C(=O)N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)