missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-nitrobenzyl Bromide 95.0+%, TCI America™

Catalog No. H06015G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H06015G Supplier TCI America Supplier No. H06015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 772-33-8
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.033
MDL Number MFCD00007340
InChI Key KFDPCYZHENQOBV-UHFFFAOYSA-N
Synonym 2-hydroxy-5-nitrobenzyl bromide, 2-bromomethyl-4-nitrophenol, alpha-bromo-4-nitro-o-cresol, phenol, 2-bromomethyl-4-nitro, koshland reagent i, 2-bromomethyl-4-nitro-phenol, 2-hydroxy-5-nitrobenzylbromide, .alpha.-bromo-4-nitro-o-cresol, o-cresol, .alpha.-bromo-4-nitro, koshland i
PubChem CID 13044
IUPAC Name 2-(bromomethyl)-4-nitrophenol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])CBr)O

Specifications

Melting Point 144°C
Color Yellow
Quantity 5 g
UN Number 1759
Formula Weight 232.03
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-nitrobenzyl Bromide