missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2'-Hydroxy-5'-nitroacetophenone 98.0+%, TCI America™
$262.13 - $947.29
Chemical Identifiers
CAS | 1450-76-6 |
---|---|
Molecular Formula | C8H7NO4 |
Molecular Weight (g/mol) | 181.147 |
MDL Number | MFCD00463816 |
InChI Key | LNCBPUWMGYOISS-UHFFFAOYSA-N |
PubChem CID | 248079 |
SMILES | CC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])O |
Chemical Identifiers
1450-76-6 | |
181.147 | |
LNCBPUWMGYOISS-UHFFFAOYSA-N | |
CC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])O |
C8H7NO4 | |
MFCD00463816 | |
248079 |
Specifications
1450-76-6 | |
C8H7NO4 | |
5 g | |
CC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])O | |
248079 | |
≥98.0% (GC,T) | |
2′-Hydroxy-5′-nitroacetophenone |
102°C | |
MFCD00463816 | |
LNCBPUWMGYOISS-UHFFFAOYSA-N | |
181.147 | |
181.15 | |
Crystalline Powder |
Safety and Handling
TSCA : No