missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methylisophthalaldehyde 98.0+%, TCI America™

Catalog No. H06835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06835G 5 g
H06831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06835G Supplier TCI America Supplier No. H06835G

Chemical Identifiers

CAS 7310-95-4
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00016596
InChI Key ZBOUXALQDLLARY-UHFFFAOYSA-N
Synonym 2-hydroxy-5-methylisophthalaldehyde, 2,6-diformyl-4-methylphenol, 1,3-benzenedicarboxaldehyde, 2-hydroxy-5-methyl, 2-hydroxy-5-methyl-1,3-benzenedicarboxaldehyde, 2-hydroxy-5-methylisophthalaldehyd, acmc-209opk, 2-hydroxy-5-methylisophthaldehyde, 2-hydroxy-5-methylisophtalaldehyde, 2-hydroxy-5-methyl-benzene-1,3-dicarbaldehyde, 5-methyl-2-hydroxy-1,3-benzenedicarboxaldehyde
PubChem CID 81744
IUPAC Name 2-hydroxy-5-methylbenzene-1,3-dicarbaldehyde
SMILES CC1=CC(=C(C(=C1)C=O)O)C=O

Specifications

Melting Point 132°C
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-methylisophthalaldehyde