CAS RN 7310-95-4
CAS RN 7310-95-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxy-5-methylisophthalaldehyde 98.0+%, TCI America™
CAS: 7310-95-4 Molecular Formula: C9H8O3 Molecular Weight (g/mol): 164.16 MDL Number: MFCD00016596 InChI Key: ZBOUXALQDLLARY-UHFFFAOYSA-N Synonym: 2-hydroxy-5-methylisophthalaldehyde,2,6-diformyl-4-methylphenol,1,3-benzenedicarboxaldehyde, 2-hydroxy-5-methyl,2-hydroxy-5-methyl-1,3-benzenedicarboxaldehyde,2-hydroxy-5-methylisophthalaldehyd,acmc-209opk,2-hydroxy-5-methylisophthaldehyde,2-hydroxy-5-methylisophtalaldehyde,2-hydroxy-5-methyl-benzene-1,3-dicarbaldehyde,5-methyl-2-hydroxy-1,3-benzenedicarboxaldehyde PubChem CID: 81744 IUPAC Name: 2-hydroxy-5-methylbenzene-1,3-dicarbaldehyde SMILES: CC1=CC(=C(C(=C1)C=O)O)C=O