missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-5-methylbenzophenone 97.0+%, TCI America™
$160.30 - $467.87
Chemical Identifiers
| CAS | 1470-57-1 |
|---|---|
| Molecular Formula | C14H12O2 |
| Molecular Weight (g/mol) | 212.248 |
| MDL Number | MFCD00002379 |
| InChI Key | OQERFUGURPLBQH-UHFFFAOYSA-N |
| Synonym | 2-hydroxy-5-methylbenzophenone, 2-benzoyl-4-methylphenol, 2-hydroxy-5-methylphenyl phenyl methanone, unii-85w615bmp7, methanone, 2-hydroxy-5-methylphenyl phenyl, 2-hydroxy-5-methyl-benzophenone, 2-hydroxy-5-methylphenyl-phenylmethanone, acmc-209cy0, dsstox_cid_27857, dsstox_rid_82610 |
| PubChem CID | 73851 |
| IUPAC Name | (2-hydroxy-5-methylphenyl)-phenylmethanone |
| SMILES | CC1=CC(=C(C=C1)O)C(=O)C2=CC=CC=C2 |
Chemical Identifiers
| 1470-57-1 | |
| 212.248 | |
| OQERFUGURPLBQH-UHFFFAOYSA-N | |
| 73851 | |
| CC1=CC(=C(C=C1)O)C(=O)C2=CC=CC=C2 |
| C14H12O2 | |
| MFCD00002379 | |
| 2-hydroxy-5-methylbenzophenone, 2-benzoyl-4-methylphenol, 2-hydroxy-5-methylphenyl phenyl methanone, unii-85w615bmp7, methanone, 2-hydroxy-5-methylphenyl phenyl, 2-hydroxy-5-methyl-benzophenone, 2-hydroxy-5-methylphenyl-phenylmethanone, acmc-209cy0, dsstox_cid_27857, dsstox_rid_82610 | |
| (2-hydroxy-5-methylphenyl)-phenylmethanone |
Specifications
| 1470-57-1 | |
| Yellow | |
| MFCD00002379 | |
| 2-hydroxy-5-methylbenzophenone, 2-benzoyl-4-methylphenol, 2-hydroxy-5-methylphenyl phenyl methanone, unii-85w615bmp7, methanone, 2-hydroxy-5-methylphenyl phenyl, 2-hydroxy-5-methyl-benzophenone, 2-hydroxy-5-methylphenyl-phenylmethanone, acmc-209cy0, dsstox_cid_27857, dsstox_rid_82610 | |
| CC1=CC(=C(C=C1)O)C(=O)C2=CC=CC=C2 | |
| 212.248 | |
| 212.25 | |
| Crystalline Powder |
| 85°C | |
| C14H12O2 | |
| 5 g | |
| OQERFUGURPLBQH-UHFFFAOYSA-N | |
| (2-hydroxy-5-methylphenyl)-phenylmethanone | |
| 73851 | |
| ≥97.0% (GC) | |
| 2-Hydroxy-5-methylbenzophenone |
Safety and Handling
TSCA : No