missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenylacetic acid, 99+%

Catalog No. AC121700250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121700250 25 g Glass bottle
AC121700050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC121700250 Supplier Thermo Scientific Chemicals Supplier No. 121700250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 621-37-4 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-37-4
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00004337
InChI Key FVMDYYGIDFPZAX-UHFFFAOYSA-N
Synonym 3-hydroxyphenylacetic acid, 2-3-hydroxyphenyl acetic acid, 3-hydroxyphenyl acetic acid, 3-hydroxybenzeneacetic acid, 3-hydroxyphenylacetate, m-hydroxyphenylacetic acid, m-hydroxyphenyl acetic acid, benzeneacetic acid, 3-hydroxy, metahydroxy phenylacetic acid, acetic acid, m-hydroxyphenyl
PubChem CID 12122
ChEBI CHEBI:17445
IUPAC Name 2-(3-hydroxyphenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(O)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 128.0°C to 131.0°C
Color Beige-Brown to White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H4CH2CO2H
Quantity 25 g
Beilstein 10, 189
Solubility Information Solubility in water: soluble in water
Formula Weight 152.15
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxyphenylacetic acid, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-684-4
TSCA TSCA

RUO – Research Use Only