missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-nitropyridine, 96%

Catalog No. AAH66852MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66852MD 250 mg
AAH6685203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66852MD Supplier Thermo Scientific Chemicals Supplier No. H66852MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4487-51-8 | C5H4N2O3 | 140.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4487-51-8
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.10
MDL Number MFCD04114149
InChI Key STJAXIFXCBWILG-UHFFFAOYSA-N
Synonym 2-hydroxy-4-nitropyridine, 4-nitropyridin-2-ol, 4-nitropyrid-2-one, 4-nitropyridin-2 1h-one, 4-nitro-2-pyridinol, 4-nitropyridine-2-ol, 4-nitro-2 1h-pyridinone, ksc497o0t, 2 1h-pyridinone, 4-nitro, 2-hydroxy-4-nitro pyridine
PubChem CID 2762823
IUPAC Name 4-nitro-1,2-dihydropyridin-2-one
SMILES [O-][N+](=O)C1=CC(=O)NC=C1

Specifications

Color Colorless to Yellow
Density 1.44 g/mL
Boiling Point 334°C
Flash Point 156°C (313°F)
Quantity 250 mg
Formula Weight 140.1
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Hydroxy-4-nitropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only