accessibility menu, dialog, popup

UserName

CAS RN 4487-51-8

IUPAC Name: 4-nitro-1,2-dihydropyridin-2-one
Synonyms: 4-nitro-1H-pyridin-2-one
Molecular Weight (g/mol): 140.1
Molecular Formula: C5H4N2O3
InChi Key: STJAXIFXCBWILG-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=O)NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results