missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-n-octyloxybenzophenone 98.0+%, TCI America™

Catalog No. H028825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H028825G 25 g
H0288500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H028825G Supplier TCI America Supplier No. H028825G

Chemical Identifiers

CAS 1843-05-6
Molecular Formula C21H26O3
Molecular Weight (g/mol) 326.436
MDL Number MFCD00027327
InChI Key QUAMTGJKVDWJEQ-UHFFFAOYSA-N
Synonym octabenzone, 2-hydroxy-4-n-octyloxybenzophenone, benzophenone-12, 2-hydroxy-4-octyloxy phenyl phenyl methanone, benzon oo, 2-hydroxy-4-octyloxy benzophenone, benzophenone 12, cyasorb uv 531, spectra-sorb uv 531, sumisorb 130
PubChem CID 15797
IUPAC Name (2-hydroxy-4-octoxyphenyl)-phenylmethanone
SMILES CCCCCCCCOC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 25 g
Formula Weight 326.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4-n-octyloxybenzophenone
EINECSNumber (4)-0141
RTECSNumber DJ1595000
TSCA Yes