missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-4-methylbenzophenone 97.0+%, TCI America™
Supplier: TCI America H157525G
Specifications
| 2-Hydroxy-4-methylbenzophenone | |
| 59°C | |
| 234°C | |
| MFCD01099035 | |
| KIZCNUWGIVQQBK-UHFFFAOYSA-N | |
| 2-benzoyl-5-methylphenol | |
| 137824 | |
| ≥97.0% (GC) |
| 3098-18-8 | |
| Yellow | |
| C14H12O2 | |
| 25 g | |
| CC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1 | |
| 212.25 | |
| 212.25 | |
| Crystalline Powder |
Chemical Identifiers
| 3098-18-8 | |
| 212.25 | |
| KIZCNUWGIVQQBK-UHFFFAOYSA-N | |
| 2-benzoyl-5-methylphenol |
| C14H12O2 | |
| MFCD01099035 | |
| 137824 | |
| CC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1 |
Safety and Handling
TSCA : No