missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-4-methylbenzophenone 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 59°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 212.25 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Hydroxy-4-methylbenzophenone |
Safety and Handling
| TSCA | No |