missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4'-methoxypropiophenone 98.0+%, TCI America™

Catalog No. H12075G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12075G Supplier TCI America Supplier No. H12075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6270-44-6
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD04037453
InChI Key MAKZPFSTXDXNJN-UHFFFAOYSA-N
PubChem CID 234836
IUPAC Name 1-(2-hydroxy-4-methoxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(OC)C=C1O

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-4′-methoxypropiophenone