accessibility menu, dialog, popup

UserName

CH3OOCH3HO

CAS RN 6270-44-6

IUPAC Name: 1-(2-hydroxy-4-methoxyphenyl)propan-1-one
Synonyms: 1-(2-hydroxy-4-methoxyphenyl)propan-1-one
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: MAKZPFSTXDXNJN-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(OC)C=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results