missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4-methoxychalcone 98.0+%, TCI America™

Catalog No. p-7131069
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14841G 1 g
H14845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14841G Supplier TCI America Supplier No. H14841G
Only null left

Chemical Identifiers

CAS 3327-24-0
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.285
MDL Number MFCD00016447
InChI Key NXBNYUSXDBHELA-DHZHZOJOSA-N
PubChem CID 5331295
IUPAC Name (E)-1-(2-hydroxyphenyl)-3-(4-methoxyphenyl)prop-2-en-1-one
SMILES COC1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2O

Specifications

Melting Point 93°C
Color Yellow
Quantity 1 g
Synonym 2′C-Hydroxy-2-(4-methoxybenzal)acetophenone, 2′C-Hydroxy-2-(4-methoxybenzylidene)acetophenone
Formula Weight 254.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-4-methoxychalcone