accessibility menu, dialog, popup

UserName

CAS RN 3327-24-0

IUPAC Name: (2E)-1-(2-hydroxyphenyl)-3-(4-methoxyphenyl)prop-2-en-1-one
Synonyms: (2E)-1-(2-hydroxyphenyl)-3-(4-methoxyphenyl)prop-2-en-1-one
Molecular Weight (g/mol): 254.29
Molecular Formula: C16H14O3
InChi Key: NXBNYUSXDBHELA-DHZHZOJOSA-N
SMILES: COC1=CC=C(\C=C\C(=O)C2=CC=CC=C2O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results